Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rosmarinic acid ≥97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (R,E)-3-(3,4-Dihydroxyphenyl)-2-((3-(3,4-dihydroxyphenyl)acryloyl)oxy)propanoic acid

H37451-03 H37451-06
AAH37451-06EA 326.62 USD InStock
AAH37451-06 AAH37451-03
Rosmarinic acid ≥97%
Rosmarinic acid


Formula: C₁₈H₁₆O₈
MW: 360.318 g/mol
Melting Pt: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00017740
CAS Number: 20283-92-5
EINECS: 606-487-1
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty