Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

H50873-MD H50873-03
AAH50873-03EA 200.99 USD InStock
AAH50873-03 AAH50873-MD
4-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine 97%
4-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine


Formula: C₁₀H₁₇N₃O
Storage Temperature: Ambient
MDL Number: MFCD08061038
CAS Number: 733748-92-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty