Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chlorophenylmagnesium bromide 1 M in MeTHF (2-Methyltetrahydrofuran)

Supplier: Thermo Scientific Chemicals
Danger

H51161-AE H51161-AD
AAH51161-AEEA 468.99 USD InStock
AAH51161-AE AAH51161-AD
3-Chlorophenylmagnesium bromide 1 M in MeTHF (2-Methyltetrahydrofuran)
3-Chlorophenylmagnesium bromide


Formula: C₆H₄BrClMg
Storage Temperature: Ambient
MDL Number: MFCD00672003
CAS Number: 36229-42-2
UN: 2924
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty