Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Propylmagnesium chloride 1 M in MeTHF (2-Methyltetrahydrofuran)

Supplier: Thermo Scientific Chemicals
Danger

H51162-AP H51162-AE
AAH51162-APEA 266.99 USD InStock
AAH51162-AP AAH51162-AE
n-Propylmagnesium chloride 1 M in MeTHF (2-Methyltetrahydrofuran)
n-Propylmagnesium chloride


Formula: C₃H₇ClMg
Storage Temperature: Ambient
MDL Number: MFCD00000474
CAS Number: 2234-82-4
UN: 3399
ADR: "4.3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty