Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Methyl-2-(tributylstannyl)imidazole 90+%

Supplier: Thermo Scientific Chemicals
Danger

H51523-03
AAH51523-03EA 275.99 USD InStock
AAH51523-03
1-Methyl-2-(tributylstannyl)imidazole 90+%
1-Methyl-2-(tributylstannyl)imidazole


Formula: C₁₆H₃₂N₂Sn
MW: 371.15
Storage Temperature: Ambient
MDL Number: MFCD01319030
CAS Number: 105494-69-7
UN: 2788
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty