Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromobenzo[c][1,2,5]oxadiazole 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Bromo-2,1,3-benzoxadiazole

H51674-03
AAH51674-03EA 55.39 USD InStock
AAH51674-03
5-Bromobenzo[c][1,2,5]oxadiazole 97%
5-Bromobenzo[c][1,2,5]oxadiazole


Formula: C₆H₃BrN₂O
MW: 199.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02682026
CAS Number: 51376-06-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty