Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Pyridin-3-yl-1H-benzimidazole 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(Pyridin-3-yl)-1H-benzo[d]imidazole

H51785-14 H51785-06
AAH51785-06EA 61.29 USD InStock
AAH51785-06 AAH51785-14
2-Pyridin-3-yl-1H-benzimidazole 97%
2-Pyridin-3-yl-1H-benzimidazole


Formula: C₁₂H₉N₃
MW: 195.23 g/mol
Melting Pt: 254…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00022665
CAS Number: 1137-67-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty