Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Boc-Phe(3-F)-OHN-Boc-3-fluoro-L-phenylalanine

H51988-MD H51988-06 H51988-03
AAH51988-03EA 95.39 USD InStock
AAH51988-03 AAH51988-MD AAH51988-06
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine 95%
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine


Formula: C₁₄H₁₈FNO₄
MW: 283.30 g/mol
Melting Pt: 75…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00672522
CAS Number: 114873-01-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty