Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Fmoc-pentafluoro-L-phenylalanineFmoc-Phe(F5)-OH

H52059-03 H52059-MD H52059-06
AAH52059-03EA 152.99 USD InStock
AAH52059-03 AAH52059-06 AAH52059-MD
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine 95%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine


Formula: C₂₄H₁₆F₅NO₄
MW: 477.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00270612
CAS Number: 205526-32-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty