Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Fmoc-3-(4-thiazolyl)-L-alanine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Fmoc-3-(4-thiazolyl)-AlanineFmoc-L-(4-thiazolyl)-Alanine

H52071-03 H52071-MD H52071-06
AAH52071-03EA 206.99 USD InStock
AAH52071-03 AAH52071-06 AAH52071-MD
N-Fmoc-3-(4-thiazolyl)-L-alanine 95%
N-Fmoc-3-(4-thiazolyl)-L-alanine


Formula: C₂₁H₁₈N₂O₄S
MW: 394.4 g/mol
Melting Pt: 178…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00672568
CAS Number: 205528-32-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty