Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-4-Benzoylphenylalanine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: H-p-Bz-Phe-OH

H52083-03 H52083-MD H52083-06
AAH52083-03EA 269.99 USD InStock
AAH52083-03 AAH52083-06 AAH52083-MD
L-4-Benzoylphenylalanine 95%
L-4-Benzoylphenylalanine


Formula: C₁₆H₁₅NO₃
MW: 269.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00238102
CAS Number: 104504-45-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty