Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Allylglycine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (S)-2-Allylglycine(S)-(-)-alpha-AllylglycineH-Gly(Ally)-OH2-Allyl-L-glycine(2S)-2-Aminopent-4-enoic acid

H52097-03 H52097-MD
AAH52097-03EA 276.99 USD InStock
AAH52097-03 AAH52097-MD
L-Allylglycine 95%
L-Allylglycine


Formula: C₅H₉NO₂
MW: 115.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002627
CAS Number: 16338-48-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty