Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-N-BOC-4-ChlorophenylalanineBoc-D-Phe(4-Cl)-OHN-Boc-4-chloro-D-phenylalanine

H52181-MD H52181-03 H52181-06
AAH52181-MDEA 27.79 USD InStock
AAH52181-MD AAH52181-06 AAH52181-03
N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine 95%
N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine


Formula: C₁₄H₁₈ClNO₄
MW: 299.75 g/mol
Melting Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00076978
CAS Number: 57292-44-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty