Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-L-β-homoaspartic acid-γ-benzyl ester 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Boc-β-HomoAsp(OBzl)-OHBoc-β-HoAsp(OBzl)-OH

H52191-MD H52191-03
AAH52191-MDEA 141.99 USD InStock
AAH52191-MD AAH52191-03
Boc-L-β-homoaspartic acid-γ-benzyl ester 95%
Boc-L-beta-homoaspartic acid-gamma-benzyl ester


Formula: C₁₇H₂₃NO₆
MW: 337.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862861
CAS Number: 254101-10-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty