Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals

H52312-MD H52312-03
AAH52312-MDEA 99.09 USD InStock
AAH52312-MD AAH52312-03
2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester 96%
2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester


Formula: C₁₃H₁₉BO₅
Storage Temperature: Ambient
MDL Number: MFCD11855980
CAS Number: 1073338-92-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty