Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: tert-Butyl 1,2,3,6-Tetrahydropyridine-1-carboxylatetert-Butyl 5,6-dihydropyridine-1(2H)-carboxylate1-Boc-1,2,3,6-tetrahydropyridine1-tert-Butoxycarbonyl-1,2,3,6-tetrahydropyridine1,2,3,6-Tetrahydropyridine-1-carboxylic acid tert-Butyl ester

H52574-MD H52574-06 H52574-03
AAH52574-06EA 325.99 USD InStock
AAH52574-06 AAH52574-MD AAH52574-03
3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester 97%
3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester


Formula: C₁₀H₁₇NO₂
MW: 183.25 g/mol
Boiling Pt: 78…84 °C/5 mm
Density: 1.029
Storage Temperature: Ambient
MDL Number: MFCD04972245
CAS Number: 85838-94-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty