Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butyl (piperidin-4-ylmethyl)carbamate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: tert-Butyl (4-Piperidylmethyl)carbamate4-(Boc-aminomethyl)piperidine4-(tert-Butoxycarbonylaminomethyl)piperidine(4-Piperidylmethyl)carbamic acid tert-Butyl ester

H52725-06 H52725-03
AAH52725-03EA 69.59 USD InStock
AAH52725-03 AAH52725-06
tert-Butyl (piperidin-4-ylmethyl)carbamate 97%
tert-Butyl (piperidin-4-ylmethyl)carbamate


Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Melting Pt: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD01631214
CAS Number: 135632-53-0
UN: 3259
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty