Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Diethylcarbamoyl)phenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(N,N-Diethylaminocarbonyl)phenylboronic acid4-(Diethylcarbamoyl)benzeneboronic acid

H52986-03 H52986-06
AAH52986-03EA 42.56 USD InStock
AAH52986-03 AAH52986-06
4-(Diethylcarbamoyl)phenylboronic acid 98%
4-(Diethylcarbamoyl)phenylboronic acid


Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
Melting Pt: 138…142 °C
Storage Temperature: Ambient
MDL Number: MFCD03411949
CAS Number: 389621-80-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty