Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Isoquinolineboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Isoquinoline-5-boronic acidIsoquinolin-5-ylboronic acid

H53230-03 H53230-MD
AAH53230-MDEA 66.89 USD InStock
AAH53230-MD AAH53230-03
5-Isoquinolineboronic acid 97%
5-Isoquinolineboronic acid


Formula: C₉H₈BNO₂
MW: 172.98 g/mol
Melting Pt: 204…206 °C
Storage Temperature: Ambient
MDL Number: MFCD03839356
CAS Number: 371766-08-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty