Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methyl-1-pentene 97%

Supplier: Thermo Scientific Chemicals
Danger

H53373-AE H53373-AC H53373-AA
AAH53373-AEEA 586.99 USD InStock
AAH53373-AE AAH53373-AA AAH53373-AC
2-Methyl-1-pentene 97%
2-Methyl-1-pentene


Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 62 °C
Melting Pt: –136 °C
Density: 0.682
Flash Pt: -27 °C(-17 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009405
CAS Number: 763-29-1
EINECS: 212-108-7
UN: 3295
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty