Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Trimethylsilyl)propiolic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(Trimethylsilyl)propynoic acid

H53380-14 H53380-03 H53380-06
AAH53380-06EA 216.99 USD InStock
AAH53380-06 AAH53380-14 AAH53380-03
3-(Trimethylsilyl)propiolic acid 97%
3-(Trimethylsilyl)propiolic acid


Formula: C₆H₁₀O₂Si
Boiling Pt: 105…110 °C/10 mm
Melting Pt: 47…49 °C
Flash Pt: 96 °C(205 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190216
CAS Number: 5683-31-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty