Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-3-formylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Chloropyridine-3-carboxaldehyde2-Chloronicotinaldehyde2-Chloro-3-pyridinecarboxaldehyde

H53399-03 H53399-14 H53399-06
AAH53399-06EA 118.99 USD InStock
AAH53399-06 AAH53399-03 AAH53399-14
2-Chloro-3-formylpyridine 98%
2-Chloro-3-formylpyridine


Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 50…54 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01315308
CAS Number: 36404-88-3
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty