Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)propargylamine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-Boc-propargylaminetert-Butyl 2-Propynylcarbamate2-Propynylcarbamic Acid tert-Butyl Ester

H53406-06 H53406-03 H53406-14
AAH53406-03EA 31.49 USD InStock
AAH53406-03 AAH53406-14 AAH53406-06
N-(tert-Butoxycarbonyl)propargylamine 97%
N-(tert-Butoxycarbonyl)propargylamine


Formula: C₈H₁₃NO₂
MW: 155.20 g/mol
Melting Pt: 40…44 °C
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD07367245
CAS Number: 92136-39-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty