Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Pentynoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Pent-2-ynoic acid

H53411-14 H53411-03 H53411-06
AAH53411-06EA 100.99 USD InStock
AAH53411-06 AAH53411-14 AAH53411-03
2-Pentynoic acid 97%
2-Pentynoic acid


Formula: C₅H₆O₂
MW: 98.10 g/mol
Melting Pt: 47…53 °C
Flash Pt: 96 °C(205 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014370
CAS Number: 5963-77-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty