Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Methyl-N-carbobenzoxy-L-glutamate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Carbobenzoxy-L-glutamic Acid 1-Methyl Ester1-Methyl N-Cbz-L-glutamateN-Cbz-L-glutamic Acid 1-Methyl EsterZ-Glu-OMe

H53417-06 H53417-03
AAH53417-06EA 438.99 USD InStock
AAH53417-06 AAH53417-03
1-Methyl-N-carbobenzoxy-L-glutamate 98%
1-Methyl-N-carbobenzoxy-L-glutamate


Formula: C₁₄H₁₇NO₆
MW: 295.29 g/mol
Melting Pt: 64…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00083278
CAS Number: 5672-83-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty