Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Phenyl-1-hexyn-3-ol 97%

Supplier: Thermo Scientific Chemicals

H53437-06 H53437-14
AAH53437-06EA 70.59 USD InStock
AAH53437-06 AAH53437-14
1-Phenyl-1-hexyn-3-ol 97%
1-Phenyl-1-hexyn-3-ol


Formula: C₁₂H₁₄O
Density: 0.949
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00046687
CAS Number: 1817-51-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty