Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Hexenyl acetate 97%

Supplier: Thermo Scientific Chemicals
Warning

H53445-22 H53445-06 H53445-14
AAH53445-14EA 115.99 USD InStock
AAH53445-14 AAH53445-06 AAH53445-22
5-Hexenyl acetate 97%
5-Hexenyl acetate


Formula: C₈H₁₄O₂
Boiling Pt: 173…174 °C
Density: 0.883
Flash Pt: 60 °C(142 °F)
Storage Temperature: Ambient
UN: 3272
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty