Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1-Dimethylpropargylamine 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Methylbut-3-yn-2-amine

H53448-06 H53448-14
AAH53448-06EA 69.99 USD InStock
AAH53448-06 AAH53448-14
1,1-Dimethylpropargylamine 95%
1,1-Dimethylpropargylamine


Formula: C₅H₉N
MW: 83.134 g/mol
Boiling Pt: 79…80 °C
Density: 0.79
Flash Pt: 2 °C(36 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008052
CAS Number: 2978-58-7
EINECS: 221-029-7
UN: 2733
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty