Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methylhippuric acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(3-Methylbenzamido)acetic acid

H53458-14 H53458-03 H53458-06
AAH53458-06EA 117.99 USD InStock
AAH53458-06 AAH53458-14 AAH53458-03
3-Methylhippuric acid 97%
3-Methylhippuric acid


Formula: C₁₀H₁₁NO₃
MW: 193.20 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
CAS Number: 27115-49-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty