Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Ethynylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Pyridylacetylene

H53465-14 H53465-06 H53465-03
AAH53465-14EA 962.99 USD InStock
AAH53465-14 AAH53465-03 AAH53465-06
3-Ethynylpyridine 97%
3-Ethynylpyridine


Formula: C₇H₅N
MW: 103.12 g/mol
Boiling Pt: 83…84 °C/30 mm
Melting Pt: 39…40 °C
Flash Pt: 57 °C(135 °F)
Storage Temperature: Ambient
MDL Number: MFCD02177459
CAS Number: 2510-23-8
UN: 1325
ADR: "4.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty