Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,13-Tetradecadiene 90+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tetradeca-1,13-diene

H53479-03 H53479-14 H53479-06
AAH53479-03EA 53.39 USD InStock
AAH53479-03 AAH53479-14 AAH53479-06
1,13-Tetradecadiene 90+%
1,13-Tetradecadiene


Formula: C₁₄H₂₆
Boiling Pt: 131 °C/17 mm
Density: 0.849
Flash Pt: 103 °C(217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014941
CAS Number: 21964-49-8
EINECS: 244-687-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty