Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopropyl(trimethylsilyl)acetylene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (Cyclopropylethynyl)trimethylsilane

H53487-06 H53487-06 H53487-14 H53487-14
AAH53487-06EA 115.99 USD InStock
CAAAH53487-14 CAAAH53487-06 AAH53487-06 AAH53487-14
Cyclopropyl(trimethylsilyl)acetylene 97%
Cyclopropyl(trimethylsilyl)acetylene


Formula: C₈H₁₄Si
MW: 138.285 g/mol
Boiling Pt: 83 °C/80 mm
Flash Pt: 25 °C(77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00671355
CAS Number: 81166-84-9
UN: 1993
ADR: "3",III
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty