Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-[Di(methylthio)methylidene]malononitrile 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(Bis(methylthio)methylene)malononitrile

H53503-14 H53503-06
AAH53503-14EA 267.99 USD InStock
AAH53503-14 AAH53503-06
2-[Di(methylthio)methylidene]malononitrile 97%
2-[Di(methylthio)methylidene]malononitrile


Formula: C₆H₆N₂S₂
MW: 170.26 g/mol
Melting Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00052730
CAS Number: 5147-80-8
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty