Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(3,5-Dibromophenyl)propionic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(3,5-Dibromophenyl)propanoic acid

H54012-03 H54012-06
AAH54012-06EA 328.99 USD InStock
AAH54012-06 AAH54012-03
3-(3,5-Dibromophenyl)propionic acid 97%
3-(3,5-Dibromophenyl)propionic acid


Formula: C₉H₈Br₂O₂
MW: 307.97 g/mol
Melting Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD11973898
CAS Number: 923977-15-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty