Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Piperidine-1-sulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

H54053-06 H54053-03 H54053-MD
AAH54053-06EA 540.99 USD InStock
AAH54053-06 AAH54053-MD AAH54053-03
Piperidine-1-sulfonyl chloride 97%
Piperidine-1-sulfonyl chloride


Formula: C₅H₁₀ClNO₂S
MW: 183.65 g/mol
Density: 1.308
Storage Temperature: Ambient
MDL Number: MFCD03250329
CAS Number: 35856-62-3
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty