Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Bromoquinolin-2(1H)-one 96%

Supplier: Thermo Scientific Chemicals
Warning

H54095-06 H54095-14 H54095-03
AAH54095-06EA 189.99 USD InStock
AAH54095-06 AAH54095-14 AAH54095-03
6-Bromoquinolin-2(1H)-one 96%
6-Bromoquinolin-2(1H)-one


Formula: C₉H₆BrNO
MW: 224.05 g/mol
Melting Pt: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD08703169
CAS Number: 1810-66-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty