Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(4-Methoxyphenyl)magnesium bromide 1 mol/l in 2-MeTHF (2-Methyltetrahydrofuran)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Anisylmagnesium bromide

H54317-AE H54317-AE
AAH54317-AEEA 168.99 USD InStock
CAAAH54317-AE AAH54317-AE
(4-Methoxyphenyl)magnesium bromide 1 mol/l in 2-MeTHF (2-Methyltetrahydrofuran)
(4-Methoxyphenyl)magnesium bromide


Formula: C₇H₇BrMgO
MW: 211.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013515
CAS Number: 13139-86-1
UN: 2924
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty