Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-N-methylbenzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N-Methyl-4-fluorobenzylamine

H54369-06 H54369-03
AAH54369-03EA 40.39 USD InStock
AAH54369-03 AAH54369-06
4-Fluoro-N-methylbenzylamine 97%
4-Fluoro-N-methylbenzylamine


Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 180…181 °C
Density: 1.056
Flash Pt: 71 °C(160 °F)
Storage Temperature: Ambient
MDL Number: MFCD03839848
CAS Number: 405-66-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty