Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-4-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridin-2-amine

H54372-MD H54372-03
AAH54372-MDEA 185.99 USD InStock
AAH54372-MD AAH54372-03
2-Amino-4-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 95%
2-Amino-4-(trifluoromethyl)-5-pyridineboronic acid pinacol ester


Formula: C₁₂H₁₆BF₃N₂O₂
MW: 288.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12923420
CAS Number: 944401-57-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty