Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Cyano-3-pyridineboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Cyanopyridine-5-boronic acid pinacol ester3-Cyanopyridine-5-boronic acid Pinacol ester5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile3-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine2-(5-Cyano-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H54385-03 H54385-MD
AAH54385-MDEA 95.59 USD InStock
AAH54385-MD AAH54385-03
5-Cyano-3-pyridineboronic acid pinacol ester 96%
5-Cyano-3-pyridineboronic acid pinacol ester


Formula: C₁₂H₁₅BN₂O₂
MW: 230.07 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD07780755
CAS Number: 402718-29-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty