Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromo-8-methoxyquinoline 96%

Supplier: Thermo Scientific Chemicals
Warning

H54386-06 H54386-MD H54386-03
AAH54386-MDEA 56.29 USD InStock
AAH54386-MD AAH54386-06 AAH54386-03
5-Bromo-8-methoxyquinoline 96%
5-Bromo-8-methoxyquinoline


Formula: C₁₀H₈BrNO
MW: 238.08 g/mol
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD04966996
CAS Number: 10522-47-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty