Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Lactose mixture of stereoisomers ∼70% beta & ∼30% alpha

Supplier: Thermo Scientific Chemicals
Synonyms: beta-D-Lactoseβ-D-Gal-(1→4)-β-D-Glc

H54447-18 H54447-30 H54447-A1
AAH54447-30EA 159.23 USD InStock
AAH54447-30 AAH54447-A1 AAH54447-18
β-Lactose mixture of stereoisomers ∼70% beta & ∼30% alpha
β-Lactose


Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064521
CAS Number: 5965-66-2
EINECS: 227-751-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty