Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

indane-5-sulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2,3-Dihydro-1H-indene-5-sulfonyl chloride2,3-Dihydro-1H-indene-5-sulphonyl chloride

H54462-MD H54462-06 H54462-06 H54462-03 H54462-03 H54462-MD
AAH54462-03EA 99.09 USD InStock
AAH54462-03 CAAAH54462-03 CAAAH54462-06 CAAAH54462-MD AAH54462-MD AAH54462-06
indane-5-sulfonyl chloride 97%
indane-5-sulfonyl chloride


Formula: C₉H₉ClO₂S
MW: 216.679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05237217
CAS Number: 52205-85-3
UN: 3261
ADR: "8",III
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty