Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Boc-(2-aminomethyl)piperidine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester2-(Aminomethyl)-1-Boc-piperidine1-Boc-2-(Aminomethyl)piperidinetert-Butyl 2-(Aminomethyl)-1-piperidinecarboxylate2-(Aminomethyl)-1-tert-butoxycarbonylpiperidine

H54507-MD H54507-06 H54507-03 H54507-03 H54507-06 H54507-MD
AAH54507-MDEA 35.39 USD InStock
CAAAH54507-03 AAH54507-MD AAH54507-06 AAH54507-03 CAAAH54507-MD CAAAH54507-06
1-Boc-(2-aminomethyl)piperidine 95%
1-Boc-(2-aminomethyl)piperidine


Formula: C11H22N2O2
MW: 214.31 g/mol
Density: 1.012
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01317791
CAS Number: 370069-31-1
UN: 3082
ADR: "9",III
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty