Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloroacetyl-2-(trifluoroacetyl)hydrazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H54647-03 H54647-06 H54647-MD
AAH54647-06EA 498.99 USD InStock
AAH54647-06 AAH54647-MD AAH54647-03
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine 97%
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine


Formula: C₄H₄ClF₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07779449
CAS Number: 762240-99-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty