Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methoxy-6-quinolineboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline

H54814-03 H54814-MD
AAH54814-MDEA 102.5 USD InStock
AAH54814-MD AAH54814-03
2-Methoxy-6-quinolineboronic acid pinacol ester 96%
2-Methoxy-6-quinolineboronic acid pinacol ester


Formula: C₁₆H₂₀BNO₃
MW: 285.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18383956
CAS Number: 1201644-36-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty