Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Amino-4-cyclopentylpiperazine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Cyclopentylpiperazin-1-amine

H54831-06 H54831-03 H54831-MD
AAH54831-MDEA 80.39 USD InStock
AAH54831-MD AAH54831-06 AAH54831-03
1-Amino-4-cyclopentylpiperazine 97%
1-Amino-4-cyclopentylpiperazine


Formula: C₉H₁₉N₃
MW: 169.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00190598
CAS Number: 61379-64-4
EINECS: 262-742-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty