Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Dimethylaminopyridinium bromide perbromide 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-(Dimethylamino)pyridine tribromide

H55010-22 H55010-14 H55010-06
AAH55010-22EA 452.88 USD InStock
AAH55010-22 AAH55010-14 AAH55010-06
4-Dimethylaminopyridinium bromide perbromide 97%
4-Dimethylaminopyridinium bromide perbromide


Formula: C₇H₁₁Br₃N₂
Melting Pt: 129…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00043439
CAS Number: 92976-81-3
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty