Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

S-Phenyl-L-cysteine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (R)-2-Amino-3-(phenylthio)propanoic acidH-Cys(phenyl)-OH

H55031-03 H55031-06 H55031-14
AAH55031-06EA 117.99 USD InStock
AAH55031-06 AAH55031-03 AAH55031-14
S-Phenyl-L-cysteine 97%
S-Phenyl-L-cysteine


Formula: C₉H₁₁NO₂S
MW: 197.252 g/mol
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01318758
CAS Number: 34317-61-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty