Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-[(Trimethylsilyl)ethynyl]benzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-((Trimethylsilyl)ethynyl)benzaldehyde

H55060-14 H55060-03 H55060-06
AAH55060-06EA 246.99 USD InStock
AAH55060-06 AAH55060-14 AAH55060-03
4-[(Trimethylsilyl)ethynyl]benzaldehyde 97%
4-[(Trimethylsilyl)ethynyl]benzaldehyde


Formula: C₁₂H₁₄OSi
MW: 202.33 g/mol
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD02093765
CAS Number: 77123-57-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty